missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10447250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10447250 1 kg 1kg
10070090 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10447250 Supplier Fisher Chemical Supplier No. C/8600/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.60
MDL Number 10981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
IUPAC Name copper(2+) sulfate
SMILES [Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 200°C
Color Gray
pH 3.5 to 4.5
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 159.6g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.