missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, Honeywell Fluka™

Product Code. 15686030 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15686030 500 g 500g
15676030 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15686030 Supplier Honeywell-Fluka Supplier No. 61230500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., anhydrous,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Quantity 500 g
Linear Formula CuSO4
UN Number UN3077
Formula Weight 159.61g/mol
Percent Purity ≥99% (RT)
Vapor Pressure 7.3 mmHg (25°C)
Density 3.603g/mL at 25°C
Chemical Name or Material Copper(II) sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.