missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) trifluoromethanesulfonate, 98%

Product Code. 11459736
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11459736

Brand: Thermo Scientific Alfa Aesar B20253.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34946-82-2 | CHCuF3O3S | 213.617 g/mol

Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) trifluoromethanesulfonate acts as a catalyst in Diels-Alder reaction and cyclopropanation reactions. It is also used as a reagent for oligosaccharide synthesis. It acts as a Lewis acid. It is also employed in the dimerization of ketone enolates and tetramethylsilane enol ethers. It is used to prepare cis vinylic sulfones and oxazoles from alkynyl sulfones and ketones respectively. Further, it plays an important role for cyclization of dienolates. In addition to this, it is useful for dehydration of alcohols.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34946-82-2
Molecular Formula CHCuF3O3S
Molecular Weight (g/mol) 213.617
MDL Number MFCD00077492
InChI Key GZWXEFRPSWBAGC-UHFFFAOYSA-N
Synonym copper; trifluoromethanesulfonic acid
PubChem CID 45051791
IUPAC Name copper;trifluoromethanesulfonic acid
SMILES C(F)(F)(F)S(=O)(=O)O.[Cu]

Specifications

Melting Point (decomposition)
Quantity 100 g
UN Number UN3261
Beilstein 4028198
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 361.68
Percent Purity 98%
Chemical Name or Material Copper(II) trifluoromethanesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.