Learn More
COPS3 Monoclonal Antibody (OTI3E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808963
Description
The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17.Specifications
| COPS3 | |
| Monoclonal | |
| Unconjugated | |
| COPS3 | |
| CSN3, SGN3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8533 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3E1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UNS2 | |
| COPS3 | |
| Human recombit protein fragment corresponding to amino acids 224-423 of human COPS3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.