Learn More
COPS3 Monoclonal Antibody (OTI3E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808963S
Description
COPS3 Monoclonal antibody specifically detects COPS3 in Human samples. It is validated for Western BlotSpecifications
| COPS3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96QF0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 117177 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3E1 | |
| Unconjugated | |
| RAB3IP | |
| COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3 | |
| Full length human recombinant protein of human RAB3IP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.