missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17453-1-AP-20UL
This item is not returnable.
View return policy
Description
Methyltransferase required for the conversion of 2-polyprenyl-6-methoxy-1,4-benzoquinol (DDMQH2) to 2-polyprenyl-3-methyl-6-methoxy-1,4-benzoquinol (DMQH2).Specifications
| COQ5 | |
| Polyclonal | |
| Unconjugated | |
| COQ5 | |
| COQ5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 52064, 84274 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5HYK3, Q9CXI0 | |
| Coq5 | |
| COQ5 Fusion Protein Ag10203 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction