Learn More
Tocris Bioscience™ CoraFluor™ 2, amine reactive
Product Code. 18074582
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074582 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18074582 Supplier Tocris Bioscience™ Supplier No. 7950/20U
Terbium cryptate FRET donor for TR-FRET assay development; amine reactive for conjugation
CoraFluor™ 2, amine reactive is a terbium-based time-resolved Förster resonance energy transfer (TR-FRET) donor for TR-FRET assay development, such as target engagement and ternary complex assays. CoraFluor™ 2 is designed with a PFP ester for reactivity with primary amines, making it suitable for labeling biomolecules, including antibodies, nanobodies, proteins and peptides. CoraFluor™ 2 is cell permeable and can be used in live cell intracellular TR-FRET assays It is exceptionally bright, providing unmatched assay sensitivity and stability in biological media over other commercially available TR-FRET donors. CoraFluor™ 2 displays red shifted excitation and is excitable by both 365 nm and 405 nm lasers, and is compatible with common acceptor dyes such as FAM/FITC, AF488, GFP and mCherry, enabling seamLess assay integration in high throughput screening applications. CoraFluor™ 1, amine reactive also available. Please refer to the protocol for further information on conjugation. CoraFluor™ is a trademark of Bio-Techne Corp.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 20 μg
Target Fluorescent Probes