missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 405 Anti-Human CD86 (BU63), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL405-65165-100TESTS
This item is not returnable.
View return policy
Description
The CD86 antibody from Proteintech is a mouse monoclonal antibody generated with b-lymphoblastoid cell line arh 77 of human CD86. This antibody recognizes human antigen. The CD86 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.Specifications
| CD86 | |
| Monoclonal | |
| 5 μl/test | |
| B7-2; B70; CD28LG2; LAB72; MGC34413 | |
| B-lymphoblastoid cell line ARH 77 | |
| 100 Tests | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| BU63 | |
| CoraLite Plus 405 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 942 | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction