missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated AR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66747-100UL
This item is not returnable.
View return policy
Description
The AR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AR. This antibody recognizes human antigen. The AR antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| AR | |
| Monoclonal | |
| 1000 μg/mL | |
| AIS; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 110-120 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1F7C12 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 367 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido