missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Lamin A/C Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

129.00€ - 349.00€

Specifications

Antigen Lamin A/C
Concentration 1000 μg/mL
Applications Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate CoraLite Plus 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234205
View Documents
Proteintech
CL488-10298-100UL
100 μL
ONLINE EXCLUSIVE PRICE
351.27€
349.00€ / 100µL
Save 2.27€
0.65% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236690
View Documents
Proteintech
CL488-10298-20UL
20 μL
ONLINE EXCLUSIVE PRICE
140.00€
129.00€ / 20µL
Save 11.00€
7% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Lamin A/C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Lamin A/C. This antibody recognizes human, mouse, rat antigen. The Lamin A/C antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

Lamin A/C
Immunocytochemistry/Immunofluorescence
CoraLite Plus 488
Rabbit
Human, Mouse, Rat
CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; HGPS; IDC; LDP1; LFP; LGMD1B; LMN1; LMNC; PRO1
IgG
Antigen affinity purification
Conjugated Antibody
1000 μg/mL
Polyclonal
Liquid
RUO
4000
Recombinant Protein
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.