missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated RAD21 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-27071-100UL
This item is not returnable.
View return policy
Description
The RAD21 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RAD21. This antibody recognizes human, canine antigen. The RAD21 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| RAD21 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5885 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| 1000 μg/mL | |
| FLJ25655; FLJ40596; HR21; HRAD21; KIAA0078; MCD1; NXP1; SCC1; hHR21 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Canine | |
| 120 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction