missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated TFAP2A/AP-2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 395.00€
Specifications
| Antigen | TFAP2A/AP-2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234843
|
Proteintech
CL488-13019-100UL |
100 μL |
450.00€ 395.00€ / 100µL Save 55.00€ 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236169
|
Proteintech
CL488-13019-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TFAP2A/AP-2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TFAP2A/AP-2. This antibody recognizes human antigen. The TFAP2A/AP-2 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| TFAP2A/AP-2 | |
| Immunocytochemistry/Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human | |
| AP-2; AP-2alpha; AP2TF; BOFS; FLJ51761; TFAP2 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 7020 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title