Learn More
Julabo™ CORIO CD-Series Heating Circulators with Open, Stainless Steel Baths and Pump Connections
BIOPHARMA
Product Code. 15450572
Change view
Click to view available options
Model:
CD-B17
CD-B39
CD-B5
Capacity (Metric):
13 to 17 L
3.5 to 5 L
35 to 41 L
Dimensions (L x W x H):
34 x 54 x 57 cm
38 x 23 x 41 cm
40 x 38 x 47 cm
Dimensions (LxWxH) Interior:
15 x 15 x 15 cm (Bath Opening)
18 x 30 x 20 cm (Bath Opening)
30 x 33 x 30 cm (Bath Opening)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Capacity (Metric) Dimensions (L x W x H) Dimensions (LxWxH) Interior Unit Size
15450572 CD-B5 3.5 to 5 L 38 x 23 x 41 cm 15 x 15 x 15 cm (Bath Opening) Each
15470572 CD-B17 13 to 17 L 40 x 38 x 47 cm 18 x 30 x 20 cm (Bath Opening) Each
15430582 CD-B39 35 to 41 L 34 x 54 x 57 cm 30 x 33 x 30 cm (Bath Opening) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15450572 Supplier Julabo™ Supplier No. 9012405
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
CORIO CD-Series Heating Circulators with Open, Stainless Steel Baths and Pump Connections are designed primarily for internal temperature control directly within the bath, however, external closed systems may be connected via the integrated pump connections.
  • Bright, White, Easy to Read Display
  • Easy to Operate Front Controls
  • Easy Switching Between Internal and External Circulation
  • Low Liquid Level Early Warning System
  • Integrated Drain Screw (except CD-B5)
  • Integrated Pump Connection (1 x M16)
  • Integrated Timer
  • PID1 and ACC Controls
  • Very Quiet
Specifications
Description Circulation Capacity Flow Rate: 15 L/min., Circulation Capacity Pressure: 0.35 bar
Capacity (Metric) 3.5 to 5 L
Flow Rate 15 L/min.
Temperature Range 20°C to 150°C (Working)
Type Heating Circulator with Open Bath
For Use With (Application) Internal and external applications at working temperatures up to +150°C
Heating Capacity 2 kW
Max. Pressure 0.35 bar
Model CD-B5
Temperature Stability ±0.03°C
Dimensions (L x W x H) 38 x 23 x 41 cm
Dimensions (LxWxH) Interior 15 x 15 x 15 cm (Bath Opening)