Learn More
Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates
Product Code. 10649541 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
White, Clear Bottom
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10649541 Black, Clear Bottom Pack of 100
10377171 White, Clear Bottom Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10649541 Supplier Corning™ Supplier No. 3651
  • ONLINE EXCLUSIVE PRICE
    1290.00€
    1205.54€ / Pack of 100
    Save 84.46€
    6% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Enhanced signal-to-noise ratio
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Black, Clear Bottom
    For Use With (Equipment) Plate readers, liquid handlers and robotic systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Temperature Range (English) 39°F to 99°F
    Temperature Range (Metric) 4°C to 37°C
    Well Color Clear