Description
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Specifications
Specifications
| Closure Size | 24-410 |
| Liner Material | PTFE |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Phenolic |
| Color | Black |
| Height (Metric) | 18.5 mm |
| No. per Pack | 144/Pk. |
| No. per Case | 144/Cs. |
| Tip Material | PTFE, Rubber |