Learn More
Corning™ Flexible Packaging - Tubing Set
Product Code. 31932505 Shop All Corning Products
Change view
Click to view available options
Product Type:
Tubing Set
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31932505 Tubing Set Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31932505 Supplier Corning™ Supplier No. 9170000
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Optional tubing sets are available in combination with all single-use bag options
36„ length 1/4„ I.D. clear chemically resistant, heat sealable, flexible tubing
Male Luer and female Luer cap
Female Luer and male Luer cap
Corning media certificates are available on our Resource Library.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tubing Set