Learn More
Corning™ MCDB 131 Media
BIOPHARMA
Product Code. 15333561
Change view
Click to view available options
Quantity:
6 x 500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15333561 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15333561 Supplier Corning™ Supplier No. 15100CV
  • ONLINE EXCLUSIVE PRICE
    309.00€
    142.32€ / Pack of 6
    Save 166.68€
    53% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured under industry-leading standards for low endotoxin content and superior results
(This is suggested replacement for 15-100-CVR.)
TRUSTED_SUSTAINABILITY
Specifications
Product Type MCDB 131 Medium
Without Additives No Glutamine
Quantity 6 x 500 mL
pH 7 to 7.6
Content And Storage 2° to 8°C