Learn More
Corning™ Flexible Packaging Systems: EVA Collection Bags
Product Code. 31932509 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
5 L
10 L
20 L
Port Detail:
M/F luers; 2 slide clamps; barb
MPC M plug/connector, barb, F luer
MPC connector; M/F luers
Male MPC connector; M/F luers
Unit Size:
Each
Pack of 15
Pack of 25
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Port Detail unitSize
31932509 20 L MPC M plug/connector, barb, F luer Pack of 15
17263574 20 L MPC M plug/connector, barb, F luer Each
15316391 5 L Male MPC connector; M/F luers Each
31932508 5 L Male MPC connector; M/F luers Pack of 25
17253574 10 L MPC M plug/connector, barb, F luer Each
31932501 10 L MPC M plug/connector, barb, F luer Pack of 15
17233574 2 L Male MPC connector; M/F luers Each
31932507 2 L Male MPC connector; M/F luers Pack of 15
17243574 20 L M/F luers; 2 slide clamps; barb Each
31932522 20 L M/F luers; 2 slide clamps; barb Pack of 15
17223574 1 L MPC connector; M/F luers Each
31932496 1 L MPC connector; M/F luers Pack of 15
15374675 10 L MPC connector; M/F luers Each
31932494 10 L MPC connector; M/F luers Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 31932509 Supplier Corning™ Supplier No. 9120039
In Stock
Add to basket
Add to basket
Meet FDA requirements as 510K Medical Device
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Order Info
The full line of bag types include: collection containers (bottom port and face port designs); tank liners (2D and 3D); cryopreservation bags; cell expansion bags; bags for HYPERStack™ vessels; and rocker cell culture bags. Collection bags, collection containers and media totes are sold individually on a per each basis.
TRUSTED_SUSTAINABILITY
Specifications
Material EVA
Sterility Sterile, Irradiated
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail MPC M plug/connector, barb, F luer
Capacity (Metric) 20 L