Learn More
Corning™ PF Media
Description
- Proprietary serum-free and protein-free growth medium that does not contain any hormones or growth factors
- Supports superior growth and viability over long-term pasasges in both adherent and suspension cell culture
- Formulated without L-glutamine
For research use only. Not for IVD use.
Specifications
Specifications
| Cell Line | CHO |
| Without Additives | No Glutamine |
| Quantity | 6 x 500 mL |