Learn More
Corning™ Plain Reusable Short Stem Plastic Funnels
Product Code. 17263554 Shop All Corning Products
Change view
Click to view available options
Diameter (Metric) Stem:
1.5 mm
4 mm
6 mm
7 mm
11 mm
15 mm
Height (Metric):
45 mm
65 mm
85 mm
110 mm
180 mm
220 mm
Length (Metric) Stem:
25 mm
35 mm
43 mm
55 mm
90 mm
95 mm
Unit Size:
Pack of 12
Pack of 24
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Stem Height (Metric) Length (Metric) Stem unitSize
17263554 1.5 mm 45 mm 25 mm Pack of 24
17283554 7 mm 85 mm 43 mm Pack of 24
17273554 4 mm 65 mm 35 mm Pack of 24
17293554 6 mm 110 mm 55 mm Pack of 12
16546834 15 mm 220 mm 95 mm Pack of 12
17253554 11 mm 180 mm 90 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17263554 Supplier Corning™ Supplier No. 6120P30
  • ONLINE EXCLUSIVE PRICE
    24.15€
    22.55€ / Pack of 24
    Save 1.60€
    6% Off
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
Durable polypropylene funnels are translucent and allow safe and easy transfers of solids or liquids from one vessel to another.
Allows safe and easy transfer of solids or liquids from one vessel to another.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Stem 1.5 mm
Material Polypropylene
Height (Metric) 45 mm
Description Constructed of Durable translucent polypropylene
Length (Metric) Stem 25 mm