Learn More
Corning™ Primaria™ Tissue Culture Flasks
GREENER_CHOICE
Product Code. 10469222 Shop All Corning Products
Click to view available options
Capacity (Metric):
50 mL
250 mL
Culture Area:
25 cm2
75 cm2
Closure Type:
Plug Seal
Vented
Neck Style:
Canted
Straight
Unit Size:
Pack of 100
Pack of 200
Product Code. 10469222
Brand: Corning™ 353810
Only null left
Add to basket
Add to basket
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Order Info
Smaller flasks packed 20 per sleeve in cases of 200 (08-772-45 in cases of 100); larger flasks packed five per sleeve in cases of 100
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Phenolic
Capacity (Metric) 250 mL
Material Polystyrene
Neck Style Straight
No. per Case 100/Cs.
No. per Pack 5/Pk.
Shape Rectangular
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented
Culture Area 75 cm2
Show More Show Less
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.