Learn More
Corning™ Primaria™ Tissue Culture Flasks
GREENER_CHOICE
Product Code. 10613672 Shop All Corning Products
Click to view available options
Capacity (Metric):
50 mL
250 mL
Culture Area:
25 cm2
75 cm2
Closure Type:
Plug Seal
Vented
Neck Style:
Canted
Straight
Unit Size:
Pack of 100
Pack of 200
Product Code. 10613672
Brand: Corning™ 353808
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Order Info
Smaller flasks packed 20 per sleeve in cases of 200 (08-772-45 in cases of 100); larger flasks packed five per sleeve in cases of 100
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Phenolic
Capacity (Metric) 50 mL
Material Polystyrene
Neck Style Canted
No. per Case 100/Cs.
No. per Pack 20/Pk.
Shape Rectangular
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented
Culture Area 25 cm2
Show More Show Less
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.