Description
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 36L Corning™ Reusable Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
Specifications
Specifications
| For Use With (Equipment) | Corning Reusable Spinner Flask (36L) |