Learn More
Corning™ Sterile Spatulas, Round end
BIOPHARMA
Product Code. 10584712 Shop All Corning Products
Change view
Click to view available options
Length (Metric):
24.76 cm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10584712 24.76 cm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10584712 Supplier Corning™ Supplier No. 3005
  • ONLINE EXCLUSIVE PRICE
    303.00€
    283.04€ / Pack of 100
    Save 19.96€
    6% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
  • Corning spatulas are designed to save time and to provide contamination-free samples.
  • Eliminates the recycling and resterilizing necessary with reusable spatulas.
  • Available in five different configurations.
  • Microspatulas available in two configurations.
  • Individually packaged.
  • Antistatic.
  • Sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Disposable Single-use
Product Type Spatula
For Use With (Application) Molecular biology applications, lyophilized samples and contamination-free sampling
Length (English) 9.75 in.
Includes Round end flat blade at one end and a larger round spoon at the other end
Material Polystyrene
Length (Metric) 24.76 cm
Sterility Sterile
Shape Spoon End, Flat Round End