Learn More
SNV Comines™ Cotton Laboratory Coats
Product Code. 12150920
Change view
Click to view available options
Size:
42
44
50
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12150920 42 Each
12160920 44 Each
12120920 50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12150920 Supplier SNV Comines™ Supplier No. ANTLB00400T1
  • ONLINE EXCLUSIVE PRICE
    27.96€
    17.00€ / Pack of 1
    Save 10.96€
    39% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Provides general light protection against splashes in the laboratory. SNV Comines™ Cotton Laboratory Coats are available in a range of sizes.
  • White in color
  • Collared with lapels
  • 4 buttons
Specifications
Product Type Xavier Lab Coat
Size 42
Gender Women's
Material Carbon
Color White
Closure Type Buttons
No. of Pockets 4
Size (Metric) Chest 92 cm
Includes 2 Flat pockets, 1 breast pocket, 1 inside pocket, opening angle
Construction 100% Cotton