Learn More
CleanSpace™ Technology Coverall
Product Code. 15940062 Shop All CleanSpace Technology Products
Change view
Click to view available options
Quantity:
25/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15940062 25/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15940062 Supplier CleanSpace™ Technology Supplier No. PAF0049
  • ONLINE EXCLUSIVE PRICE
    61.93€
    48.00€ / Pack of 1
    Save 13.93€
    22% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Disposable coverall protects CleanSpace™ respirators from dirt and splash.
  • Used with the revolutionary CleanSpace™- a light weight PAPR with no hoses/belts
  • Materials: Spun polymer fibres
  • Easy and quickly fitted and removed from the power unit
  • Compatible with all CleanSpace™ Respirators
Specifications
Disposable Reusable
Product Type Blower Cover
For Use With (Equipment) Standard and Large Case Filters
Model PAF-0049
Quantity 25/Pk.