missing translation for 'onlineSavingsMsg'
Learn More

CP 1282 (>85%), TRC

Product Code. 30362715
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362715 10 mg 10mg
30361791 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30362715 Supplier Toronto Research Chemicals Supplier No. TRCC78110010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69712-30-7
Molecular Formula C17H17N7O8S4
Molecular Weight (g/mol) 575.62
Synonym (6R-cis)-7-[[[(4-Carboxy-2,3-dihydro-3-oxo-5-isothiazolyl)thio]acetyl]amino]-7-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, (6R,7S)-7-[[[(4-Carboxy-2,3-dihydro-3-oxo-5-isothiazolyl)thio]acetyl]amino]-7-methoxy-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
IUPAC Name (6R,7S)-7-[[2-[(4-carboxy-3-oxo-1,2-thiazol-5-yl)sulfanyl]acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CN1C(=NN=N1)SCC2=C(N3[C@@H]([C@@](C3=O)(NC(=O)CSC4=C(C(=O)NS4)C(=O)O)OC)SC2)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H17N7O8S4/c1-23-16(19-21-22-23)35-4-6-3-34-15-17(32-2,14(31)24(15)9(6)12(29)30)18-7(25)5-33-13-8(11(27)28)10(26)20-36-13/h15H,3-5H2,1-2H3,(H,18,25)(H,20,26)(H,27,28)(H,29,30)/t15-,17+/m1/s1
Formula Weight 575.0021
Chemical Name or Material CP 1282
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.