missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CP 94253 hydrochloride
GREENER_CHOICE

Product Code. 18044412
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18044412

Brand: Tocris Bioscience™ 1317/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective 5-HT1B agonist

Potent, selective 5-HT1B agonist (Ki values are 89, 2, 860, 49 and 1,600 nM for 5-HT1A, 5-HT1B, 5-HT1C, 5-HT1D and 5-HT2 receptors respectively). Centrally active upon systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845861-39-4
Molecular Formula C15H20ClN3O

Specifications

Quantity 50 mg
Formula Weight 293.8
Purity >98%
Molecular Weight (g/mol) 293.795
Chemical Name or Material CP 94253 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.