missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPA1 (Carboxypeptidase A1) Monoclonal Antibody (OTI1D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00€ - 540.87€
Specifications
| Antigen | CPA1 (Carboxypeptidase A1) |
|---|---|
| Clone | OTI1D11 |
| Concentration | 0.87 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17591812
|
Origene Technologies
TA504520S |
30 μL |
242.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17571812
|
Origene Technologies
TA504520 |
100 μL |
540.87€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CPA1 (Carboxypeptidase A1) Monoclonal antibody specifically detects CPA1 (Carboxypeptidase A1) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CPA1 (Carboxypeptidase A1) | |
| 0.87 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| 1357 | |
| Primary |
| OTI1D11 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P15085 | |
| IgG2a | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title