missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 1/4 in. Flow Size

Product Code. 30968795
Change view
Click to view available options
For Use With:
N/A
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
30968795 N/A Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30968795 Supplier CPC™ Supplier No. MPC30003MB25
  • ONLINE EXCLUSIVE PRICE
    75.00€
    70.43€ / Pack of 25
    Save 4.57€
    6% Off
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Detach these fittings with a simple press of the thumb latch. Cole-Parmer™ CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Flow Rate: Cv∽5.5 (79.25) max.

  

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials
  • O-Ring: Silicone Seal USP Class VI

  

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

 

Specifications

Certifications/Compliance USP Class VI, ADCF
Autoclavable Autoclavable
Material Polycarbonate
Diameter (Metric) Outer 19.1 mm
Overall Length (Metric) 31.5 mm
For Use With N/A
Min. Operating Temperature (Metric) -40°C
Color Purple
Max. Operating Temperature (Metric) 121° C
Product Type Quick Disconnect Fitting
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.