missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20249-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is expected to specifically recognize CPLX2 but not other complexins.
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.Specifications
| CPLX2 | |
| Polyclonal | |
| Unconjugated | |
| CPLX2 | |
| 921 L, complexin 2, Complexin II, CPLX2, CPX 2, CPX II, CPX2, Hfb1, Synaphin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10814, 116657, 12890 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P84086, P84087, Q6PUV4 | |
| CPLX2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction