missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Antibody (CPS1/8416), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21108JF549
583.20 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
CPS1 Monoclonal antibody specifically detects CPS1 in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
CPS1 | |
Monoclonal | |
Janelia Fluor 549 | |
carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 | |
Recombinant human CPS1 protein | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
1373 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
CPS1/8416 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction