missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15184-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can bind the close sequences genes.
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable), together with a carnitine-acylcarnitine translocase. CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| CPT1A | |
| Polyclonal | |
| Unconjugated | |
| CPT1A | |
| CPT I, CPT1, CPT1 L, CPT1A, CPTI L, L CPT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12894, 1374, 25757 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32198, P50416, P97742 | |
| Cpt1a | |
| CPT1A Fusion Protein Ag7202 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction