Learn More
WTW™ CR 3200 and CR 4200 Thermoreactors Cables
Product Code. 11751546
Change view
Click to view available options
Product Type:
PC Connection Cable
Printer Connection Cable
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11751546 PC Connection Cable Each
11741546 Printer Connection Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11751546 Supplier WTW™ Supplier No. 250102
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WTW offers printers with direct data output via a RS232 interface as well as suitable cables for the communication between devices and the PC
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CR 3200 and CR 4200 thermo reactors
Quantity Ea.
Product Type PC Connection Cable