Learn More
Abeba™ Crawler ALU 34580 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand: Abeba™ 34580/45
Additional Details : Weight : 1.23000kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Smooth leather white/blue
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Discontinued model
Specifications
CE EN ISO 20345:2011 S1 SRC | |
Leather | |
Slip-resistant | |
34580 |
White | |
45 | |
Polyurethane | |
Aluminum |