Learn More
Abeba™ Crawler ALU 34580 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand:  Abeba™ 34580/46
Additional Details : Weight : 1.23000kg
 View more versions of this product
Product Code. 15660018
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather white/blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Leather
Slip-resistant
34580
White
46
Polyurethane
Aluminum