Learn More
CRB1 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26598-1-AP-20UL
 View more versions of this product
Product Code. 17581346
  • 145.00€ / 20µL
Description
The CRB1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CRB1. This antibody recognizes mouse antigen. The CRB1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
CRB1
Polyclonal
Rabbit
Primary
Mouse
Immunohistochemistry
Unconjugated
20 μL
23418