missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Creatine Kinase MB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA513830
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, porcine and rabbit based on sequence homology.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
Specifications
| Creatine Kinase MB | |
| Polyclonal | |
| Unconjugated | |
| Ckb | |
| Bck; B-CK; Brain creatine kinase; CK MB; Ck3; Ck-3; CKB; Ckbb; Ckbr; Ckm; CKMM; CPK-B; CPK-M; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, M-type; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29; MCK; M-CK | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 1152, 1158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P06732, P12277 | |
| Ckb, CKM | |
| KLH conjugated synthetic peptide between 283-313 amino acids from the C-terminal region of human CKM. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction