missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRELD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27017-1-AP-20UL
This item is not returnable.
View return policy
Description
CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.Specifications
| CRELD2 | |
| Polyclonal | |
| Unconjugated | |
| CRELD2 | |
| CRELD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79174 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6UXH1 | |
| CRELD2 | |
| CRELD2 Fusion Protein Ag25553 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction