Learn More
Invitrogen™ Crenezumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559073
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
DLX212 is a biosimilar to anti-human amyloid beta antibodies, specifically designed for research applications such as ELISA and functional assays. DLX212 targets the beta amyloid peptide, with its specificity stemming from its recombinant monoclonal antibody structure, offering researchers a reliable tool in in vivo studies. Its application is strictly within the realm of research, aiding studies focused on amyloid pathologies, primarily Alzheimer's disease. DLX212 serves as an alternative to other anti-beta amyloid antibodies like crenezumab, with its preparation allowing it to be versatile across multiple assay platforms.
Specifications
| DLX212 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MABT5102A; RG7412 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.