missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRISP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148881
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
May have a role in sperm-egg fusion and maturation.
Specifications
| CRISP1 | |
| Polyclonal | |
| Unconjugated | |
| CRISP1 | |
| Acidic epididymal glycoprotein homolog; acidic epididymal glycoprotein-like 1; AEGL1; AEG-like protein; AEG-related protein; ARP; CRISP1; CRISP-1; cysteine rich secretory protein 1; cysteine-rich secretory protein 1; cysteine-rich secretory protein-1 delta; epididymis secretory protein Li 57; HEL-S-57; HSCRISP1D; HSCRISP1G; HUMARP | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 11571 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q03401 | |
| CRISP1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CRISP-1, Gln20-His244. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction