missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPLD2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00083716-B01P
This item is not returnable.
View return policy
Description
CRISPLD2 Polyclonal antibody specifically detects CRISPLD2 in Human samples. It is validated for Western Blot
Specifications
| CRISPLD2 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| CRISP11, CRISP-11, Cysteine-rich secretory protein 11, cysteine-rich secretory protein LCCL domain containing 2, cysteine-rich secretory protein LCCL domain-containing 2, DKFZP434B044, LCCL domain containing cysteine-rich secretory protein 2, LCCL domain-containing cysteine-rich secretory protein 2, LCRISP2, MGC74865 | |
| CRISPLD2 (AAH63012.1, 1 a.a. - 449 a.a.) full-length human protein. MSCVLGGVIPLGLLFLVCGSQGYLLPNVTLLEELLSKYQHNESHSRVRRAIPREDKEEILMLHNKLRGQVQPQASNMEYMTWDDELEKSAAAWASQCIWEHGPTGLLVSIGQNLGAHWGRYRSPGFHVQSWYDEVKDYTYPYPSECNPWCPERCSGPMCTHYTQIVWATTNKIGCAVNTCRKMTVWGEVWENAVYFVCNYSPKGNWIGEAPYKNGRPCSECPPSYGGSCRNNLCYREETYTPKPETDEMNEVETAPIPEENHVWLQPRVMRPTKPKKTSAVNYMTQVVRCDTKMKDRCKGSTCNRYQCPAGCLNHKAKIFGSLFYESSSSICRAAIHYGILDDKGGLVDITRNGKVPFFVKSERHGVQSLSKYKPSSSFMVSKVKVQDLDCYTTVAQLCPFEKPATHCPRIHCPAHCKDEPSYWAPVFGTNIYADVSRMHFQQLSRKAC | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAH63012.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 83716 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction