missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | CRMP2 |
|---|---|
| Concentration | 0.64 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803454
|
Proteintech
14686-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893444
|
Proteintech
14686-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.Specifications
| CRMP2 | |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| O08553, P47942, Q16555 | |
| 12934, 1808, 25416 | |
| CRMP2 Fusion Protein Ag5985 | |
| Primary | |
| -20°C | |
| DPYSL2 |
| 0.64 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CRMP 2, CRMP2, DHPRP2, dihydropyrimidinase like 2, DPYSL2, DRP 2, DRP2, N2A3, ULIP 2, ULIP2, Unc 33 like phosphoprotein 2 | |
| DPYSL2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title