Learn More
Mecmesin™ Test Hook
Product Code. 11917989
Encompass
Change view
Click to view available options
Connections:
10-32 UNF female thread
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections Unit Size
11917989 10-32 UNF female thread Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11917989 Supplier Mecmesin™ Supplier No. 432120
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Medium Fixed Hook fitted with 10-32 UNF female thread. Used for general purpose tension tests.
Specifications
Product Type Test Hook
Connections 10-32 UNF female thread
For Use With (Application) General purpose tension tests