Learn More
Bochem™ Crucible Tongs
Product Code. 10376141
Change view
Click to view available options
Length (Metric):
220 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10376141 220 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10376141 Supplier Bochem™ Supplier No. 2141
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Biochem™ Crucible Tongs - Scissor-like tools with concave pincers that allow the user to grasp a hot crucible, flask, evaporating dish or small beaker.
  • Crucible handling
  • Rugged type
  • Made from electrolytically polished stainless steel.
Specifications
For Use With (Application) Handling Hot Crucibles, Flasks, Evaporating Dishes, Small Flasks
Tip Style Concave Pincers
Length (Metric) 220 mm
Material Stainless Steel (18/10)
Type Crucible Tongs