Learn More
Sperian™ Honeywell™ Cryogenic Gloves
Product Code. 11876920
Change view
Click to view available options
Size:
9
10
11
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11876920 11 Each
12201448 9 Each
12201458 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11876920 Supplier Sperian™ Supplier No. 205868511
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Provides protection against small drops of liquid gas up to -170°C. Sperian™ Honeywell™ Cryogenic Gloves are fully lined with 2 aluminum/fleece layers.
  • Water-repellant silicone cowhide grain leather
  • Gunn cut, winged thumb, crispin style
  • Reinforcement between thumb and index finger
  • 200mm split leather cuff
  • Velcro™ adjustment strap on side of the palm
  • Cuff offers good forearm protection against mechanical hazards and limits any risk of intrusion
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryogenic Gloves
Size 11
Thickness (Metric) 1.1/1.3 mm
Resistant To Resistance to conductive cold: 2, Resistance to contact cold: 2
Tear Rating 2
Length (Metric) 400 mm