Learn More
STEMCELL Technologies CryoStor™ CS5
Description
- CryoStor™ CS5 is a uniquely formulated serum-free, animal component-free and defined cryopreservation medium containing 5% dimethyl sulfoxide (DMSO)
- Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS5 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
- CryoStor™ CS5 is cGMP-manufactured with USP grade components
Tekniske data
Tekniske data
| Cell Type | CHO Cells, Mesenchymal Stem and Progenitor Cells, Other, Pluripotent Stem Cells |
| For Use With (Application) | Cryopreservation |
| Quantity | 5 x 10 mL |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?