Learn More
Epredia™ Cryotome Cryocassettes
Product Code. 12698036 Shop All Epredia Products
Click to view available options
:
Variety pack (one each of 77210150 through 77210153)
Product Code. 12698036
Brand: Epredia™ 77210154
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use Epredia™ Cryotome™ Cryocassettes with FSE/FE and SME Cryotomes™.
Product Note:
  • The Cryotome FSE Cryostat has specimen holder components pre-mounted in cryochamber (cryocassette clamp, XY orienting head, and Peltier). These are replaceable only through manufacturer service.
TRUSTED_SUSTAINABILITY
Specifications
Description Variety pack (one each)
For Use With (Equipment) FSE/FE/SME