Learn More
BD Biosciences™ CS&T Research Beads for use with BD FACSDiva™ Version 7 or Later (RUO)
Product Code. 15836329 Shop All BD Biosciences Products
Change view
Click to view available options
No. of Tests:
150 Tests
50 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests Unit Size
15836329 50 Tests Each
15846329 150 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15836329 Supplier BD Biosciences™ Supplier No. 655050
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Automatically characterize, track, and report measurements of supported BD digital flow cytometers.
  • Designed for use on BD™ flow cytometers running BD FACSuite™ software version 7.0 or later.
  • FC beads enable the software to automatically characterize, track, and report measurements of supported BD digital flow cytometers.
  • CS&T research beads are dyed with fluorochromes that are excited by the cytometer's lasers.
  • Provides cost efficiency with low per-test cost and multiple kit configurations.
  • Produced under GMP for improved quality and reproducibility.
  • Enhances the cytometer setup workflow, supporting over 100 laser and filter combinations.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store vials at 2°C–8°C and protect from light. Do not freeze.
No. of Tests 50 Tests
Stability The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 24 hours at 2°C–8°CC or 8 hours at 15°C–25°C when protected from light
Product Type Fluorescent Polystyrene Beads
Product Title
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.