missing translation for 'onlineSavingsMsg'
Learn More

CSNK1A1/CSNK1A1L Antibody (PSH04-58), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-33146

Product Code. 30149041

  • 496.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CSNK1A1/CSNK1A1L Monoclonal antibody specifically detects CSNK1A1/CSNK1A1L in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CSNK1A1/CSNK1A1L
Monoclonal
1 mg/mL
Western Blot 1:2000, Flow Cytometry 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1-2ug/sample, Immunohistochemistry-Paraffin 1:1000
casein kinase I isoform alpha, casein kinase I isoform alpha-like, CK1, CKI-alpha-like
Recombinant protein within human CSNK1A1 aa 1-337/ 337. (Uniprot: P48729)
100 μL
Primary
Human, Mouse, Rat
Purified
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
PSH04-58
Unconjugated
PBS (pH7.4), 0.1% BSA and 40% Glycerol
Rabbit
Protein A purified
RUO
1452
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.